Skip to Content
Merck
All Photos(1)

Documents

8.20246

Sigma-Aldrich

2-tert-Butyl-4-methoxyphenol

for synthesis

Synonym(s):

2-tert-Butyl-4-methoxyphenol, BHA, 3-tert-Butyl-4-hydroxyanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)3C]-4-(CH3O)C6H3OH
CAS Number:
Molecular Weight:
180.24
MDL number:
UNSPSC Code:
12352104
EC Index Number:
246-563-8
NACRES:
NA.22

vapor pressure

1.33 hPa ( 105 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

880 mg/kg LD50, oral (Rat)

pH

4.8 (25 °C, 10 g/L in H2O)

bp

263 °C/1013 hPa

mp

48-63 °C

transition temp

flash point 142.1 °C

storage temp.

2-30°C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Application

<ul>
<li><strong><strong>Utilization in solvatochromic dye systems:</strong></strong> 2-tert-Butyl-4-methoxyphenol is used to synthesize 2-(tert-butyl)-4-methoxy-6-(naphthalen-1-yldiazenyl)phenol) with diazotized naphthylamine as diazo component and also is used to produce a novel azo compound (Uzan, 2023).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service