Skip to Content
Merck
All Photos(1)

Documents

567440

Millipore

Sodium chloride

ACS Reagent Grade.

Synonym(s):

NaCl, Halite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

grade

ACS reagent

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.005% Insoluble material

color

white

mp

801 °C (lit.)

solubility

water: soluble

cation traces

heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

ACS Reagent Grade.
ACS Reagent Grade. Has an optimal pH of 5.0-9.0

Application


  • Enhanced versus standard hydration in acute ischaemic stroke: REVIVE - a randomized clinical trial.: This clinical trial investigates the efficacy of sodium chloride in enhanced hydration protocols for patients with acute ischemic stroke, aiming to improve clinical outcomes. (Lin et al., 2024).

  • A Coalescing Filter for Liquid-Liquid Separation and Multistage Extraction in Continuous-Flow Chemistry.: Sodium chloride is utilized in developing a coalescing filter for efficient liquid-liquid separation and extraction processes in continuous-flow chemistry. (Daglish et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 3 years at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service