Skip to Content
MilliporeSigma

Skip To

1.17828

Peroxidase Test Strips, colorimetric

greener alternative

MQuant®

Synonym(s):

Peroxidase chemical test strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 tests
Please contact Customer Service for Availability
$137.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$137.00


Please contact Customer Service for Availability

Request a Custom OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Peroxidase Test, colorimetric, MQuant®

description

semi-quantitative

Quality Segment

product line

MQuant®

specific analyte(s)

peroxidase

reaction suitability

reaction type: enzymatic reaction

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

food testing
milk and dairy products

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric with test strips MQuant®

The product quality of milk depends heavily on successful pasteurization. To verify this, the lactoperoxidase activity must be measured regularly. The MQuant® Peroxidase test makes process control during milk pasteurization easier, without incurring high costs. Under the catalytic effect of peroxidase, an organic redox indicator is converted into a blue compound. The visual comparison of the reaction zone of the test strip with the fields of a color scale shows whether there is any peroxidase activity present (+, ++) or not (-).

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.This product has benefited from:
  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.100111.167311.10081
detection method

colorimetric

detection method

colorimetric

detection method

reflectometric

detection method

colorimetric

specific analyte(s)

peroxidase

specific analyte(s)

hydrogen peroxide

specific analyte(s)

hydrogen peroxide

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: enzymatic reaction

reaction suitability

reaction type: enzymatic reaction, reagent type: oxidant

reaction suitability

reaction type: enzymatic reaction, reagent type: oxidant

reaction suitability

reaction type: enzymatic reaction, reagent type: oxidant

product line

MQuant®

product line

MQuant®

product line

Reflectoquant®

product line

MQuant®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

application(s)

food testing
milk and dairy products

application(s)

disinfection control
milk and dairy products
swimming pools
wastewater

application(s)

disinfection control

application(s)

disinfection control
milk and dairy products
swimming pools
wastewater


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters