Skip to Content
Merck
All Photos(4)

Key Documents

1.13681

Millipore

Anaerobic jar

For creating an oxygen free or oxygen depleted environment, 2.5 Liters

Synonym(s):

Oxygen free vessel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

Quality Level

feature

Used for creating an oxygen free or oxygen depleted environment.

manufacturer/tradename

Anaerocult

application(s)

microbiology

compatibility

ANAEROTEST

General description

The anaerobic jar consists of two functional parts: a lid and a base. The jar is manufactured using high-quality materials, ensuring that it is leak-proof and that the lid always fits properly. The anaerobic jar is to be used together with the Petri dish rack (1.07040). The plate basket helps to conveniently place the plates into the jar and comes with an integrated holding clip for the Anaerotest strip.

Application

Anaerobic jar is used for cultivation of anaerobic and microaerophilic microorganisms in defined atmospheric conditions.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service