Skip to Content
Merck
All Photos(1)

Documents

1.00473

Supelco

Water

for inorganic trace analysis Suprapur®

Synonym(s):

DI water, Deionized water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21

grade

for inorganic trace analysis

Quality Level

vapor pressure

23 hPa ( 20 °C)

form

liquid

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Ag (Silver): ≤ 1.0 ppb
Al (Aluminium): ≤ 1.0 ppb
As (Arsenic): ≤ 1.0 ppb
Au (Gold): ≤ 1.0 ppb
B (Boron): ≤ 1.0 ppb
Ba (Barium): ≤ 1.0 ppb
Be (Beryllium): ≤ 1.0 ppb
Bi (Bismuth): ≤ 1.0 ppb
Ca (Calcium): ≤ 1.0 ppb
Cd (Cadmium): ≤ 1.0 ppb
Co (Cobalt): ≤ 1.0 ppb
Cr (Chromium): ≤ 1.0 ppb
Cu (Copper): ≤ 1.0 ppb
Fe (Iron): ≤ 1.0 ppb
Ga (Gallium): ≤ 1.0 ppb
Ge (Germanium): ≤ 1.0 ppb
Hg (Mercury): ≤ 1.0 ppb
In (Indium): ≤ 1.0 ppb
K (Potassium): ≤ 1.0 ppb
Li (Lithium): ≤ 1.0 ppb
Mg (Magnesium): ≤ 1.0 ppb
Mn (Manganese): ≤ 1.0 ppb
Mo (Molybdenum): ≤ 1.0 ppb
Na (Sodium): ≤ 1.0 ppb
Ni (Nickel): ≤ 1.0 ppb
Pb (Lead): ≤ 1.0 ppb
Pd (Palladium): ≤ 1.0 ppb
Pt (Platinum): ≤ 1.0 ppb
Sb (Antimony): ≤ 1.0 ppb
Sn (Tin): ≤ 1.0 ppb
Sr (Strontium): ≤ 1.0 ppb
Ti (Titanium): ≤ 1.0 ppb
Tl (Thallium): ≤ 1.0 ppb
V (Vanadium): ≤ 1.0 ppb
Zn (Zinc): ≤ 1.0 ppb
Zr (Zirconium): ≤ 1.0 ppb
Actual analysis values are subject to unavoidable systematic variations in this concentration range.

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service