Skip to Content
Merck
All Photos(1)

Key Documents

06-676

Sigma-Aldrich

Anti-mGluR2/3 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GLUR2, Anti-MGLUR2, Anti-mGlu2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

rat ... Grm2(24415)

Specificity

mGluR2 and mGluR3

Immunogen

peptide (KQFVPTVCNGREVVDSTTSSL) corresponding to amino acids 853-872 of human metabotropic glutamate receptor 2

Application

Anti-mGluR2/3 Antibody is an antibody against mGluR2/3 for use in IH, IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on rat brain microsomal preparation

Target description

100-110kDa

Physical form

0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Behnam Ghasemzadeh et al.
Pharmacology, biochemistry, and behavior, 92(3), 383-392 (2009-01-13)
Glutamatergic neurotransmission plays an important role in the behavioral and molecular plasticity observed in cocaine mediated locomotor sensitization. Recent studies show that glutamatergic signaling is regulated by receptor trafficking, synaptic localization, and association with scaffolding proteins. The trafficking of the
Differential presynaptic localization of metabotropic glutamate receptor subtypes in the rat hippocampus.
Shigemoto, R, et al.
The Journal of Neuroscience, 17, 7503-7522 (1997)
Yoshiaki Miyamoto et al.
The international journal of neuropsychopharmacology, 20(12), 1027-1035 (2017-10-12)
Several clinical studies have suggested that N-acetylaspartate and N-acetylaspartylglutamate levels in the human brain are associated with various psychiatric disorders, including major depressive disorder. We have previously identified Shati/Nat8l, an N-acetyltransferase, in the brain using an animal model of psychosis.
L Emile et al.
Neuropharmacology, 35(5), 523-530 (1996-05-01)
Glutamic acid is the major excitatory amino acid of the central nervous system which interacts with two receptor families, the ionotropic and metabotropic glutamate receptors. The metabotropic glutamate receptors (mGluRs) are coupled to G proteins and can be divided into
Design and synthesis of systemically active metabotropic glutamate subtype-2 and -3 (mGlu2/3) receptor positive allosteric modulators (PAMs): pharmacological characterization and assessment in a rat model of cocaine dependence.
Dhanya, RP; Sheffler, DJ; Dahl, R; Davis, M; Lee, PS; Yang, L; Nickols, HH; Cho et al.
Journal of medicinal chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service