Skip to Content
Merck
All Photos(1)

Key Documents

05-636-AF488

Sigma-Aldrich

Anti-phospho Histone H2A.X (Ser139), clone JBW301, Alexa Fluor 488 Conjugate Antibody

clone JBW301, from mouse, ALEXA FLUOR 488

Synonym(s):

Histone H2AX, H2a/x, Histone H2A.X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

JBW301, monoclonal

species reactivity

human

species reactivity (predicted by homology)

vertebrates (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer139)

Gene Information

human ... H2AX(3014)

General description

Histone H2A.X is a variant of histone H2A, and is similarly associated with genomic DNA. However, histone is structurally different from other members of the H2A family in possessing a C-terminal tail that contains the Ser139 residue that is phosphorylated in response to breaks in double-stranded DNA. The phosphorylation of H2A.X is a rapid process that is mediated by ATM/ATR proteins.

Application

This Anti-phospho Histone H2A.X (Ser139) antibody, clone JBW301, Alexa Fluor 488 Conjugate is validated for use in ICC for the detection of phospho Histone H2A.X (Ser139).

Quality

Evaluated by Immunocytochemistry in untreated (negative control) and Staurosporin treated HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Histone H2A.X (Ser139) in Staurosporin treated HeLa cells, and not in untreated HeLa cells.

Target description

17 kDa calculated

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lara H El Touny et al.
Oncotarget, 12(21), 2114-2130 (2021-10-23)
The therapeutic efficacy of temozolomide (TMZ) is hindered by inherent and acquired resistance. Biomarkers such as MGMT expression and MMR proficiency are used as predictors of response. However, not all MGMTlow/-ve/MMRproficient patients benefit from TMZ treatment, indicating a need for
Lihua H Wang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(3), 1073-1084 (2010-01-28)
Circulating tumor cells (CTC) in peripheral blood of patients potentially represent a fraction of solid tumor cells available for more frequent pharmacodynamic assessment of drug action than is possible using tumor biopsy. However, currently available CTC assays are limited to
Huan Zhang et al.
Frontiers in oncology, 12, 999302-999302 (2022-12-17)
Poly(ADP-ribose) (PAR) polymerase inhibitors (PARPi) either have been approved or being tested in the clinic for the treatment of a variety of cancers with homologous recombination deficiency (HRD). However, cancer cells can develop resistance to PARPi drugs through various mechanisms
Jolien Van den Bossche et al.
Cancers, 11(12) (2019-12-05)
Polo-like kinase 1 (Plk1), a master regulator of mitotic cell division, is highly expressed in non-small cell lung cancer (NSCLC) making it an interesting drug target. We examined the in vitro therapeutic effects of volasertib, a Plk1 inhibitor, in combination
Lidia F Hernandez et al.
Cell death discovery, 7(1), 134-134 (2021-06-06)
Ovarian cancer is the most lethal gynecological cancer in the US. Standard treatment consists of surgery followed by chemotherapies relying on apoptotic tumor cell death. Most women with advanced stage disease will relapse, suggesting that this disease is characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service