Skip to Content
Merck
All Photos(2)

Key Documents

05-100-I

Sigma-Aldrich

Anti-HDAC1 Antibody, clone 2E10

clone 2E10, Upstate®, from mouse

Synonym(s):

HDAC1, Histone deacetylase 1, HD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E10, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC1(3065)
mouse ... Hdac1(433759)

General description

Histone deacetylase 1 is a nuclear enzyme encoded by the Hdac1 gene (also known as HD1, RPD3, MommeD5, Hdac1-ps) in mouse and HDAC1 (also known as GoON-10, RPD3L1) gene in human. Hdac1 plays an important role in epigenetic regulation of gene expression by catalyzing the hydrolysis of N(6)-acetylated histone lysine residues. Hdac1 is also known to deacetylate p53 and thereby downregulate p53 activity.

Immunogen

KLH-conjugated, synthetic peptide (C-EEKPEAKGVKEEVKLA) corresponding to amino acids 467-482 of mouse HDAC1. The immunizing sequence is identical in human.

Application

Anti-HDAC1 Antibody, clone 2E10 is an antibody against HDAC1 for use in Western Blotting, Immunoprecipitation.
Immunoprecipitation Analysis A representative lot was used to immunoprecipitate HDAC1 from HeLa cell lysate, followed by immune complex enzyme activity assay using HDAC Assay Kit (Fluorometric Detection), Cat. No. 17-356.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected HDAC1 in 10 µg of A431 cell lysate.

Target description

~55 kDa observed

Linkage

Replaces: 05-100

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Rivas et al.
Cellular and molecular gastroenterology and hepatology, 12(5), 1669-1682 (2021-07-11)
Epigenetic regulation of gene expression plays a critical role in the development of liver cancer; however, the molecular mechanisms of epigenetic-driven liver cancers are not well understood. The aims of this study were to examine molecular mechanisms that cause the
Tarin M Bigley et al.
Journal of virology, 87(13), 7393-7408 (2013-04-26)
Human cytomegalovirus (HCMV) is a common agent of congenital infection and causes severe disease in immunocompromised patients. Current approved therapies focus on inhibiting viral DNA replication. The HCMV kinase pUL97 contributes to multiple stages of viral infection including DNA replication
Betina González et al.
Frontiers in cell and developmental biology, 8, 216-216 (2020-04-23)
Paternal environmental perturbations, including cocaine intake, can affect the development and behavior of the offspring through epigenetic inheritance. However, the mechanism by which cocaine alters the male germ cells epigenome is almost unexplored. Here, we report that cocaine-treated male mice
Alok Jha et al.
Cells, 9(4) (2020-04-23)
Apolipoprotein L1 (APOL1)-miR193a axis has been reported to play a role in the maintenance of podocyte homeostasis. In the present study, we analyzed transcription factors relevant to miR193a in human podocytes and their effects on podocytes' molecular phenotype. The motif
Arturo Mendoza et al.
Science signaling, 14(709), eabh3839-eabh3839 (2021-11-17)
[Figure: see text].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service