Skip to Content
Merck
All Photos(1)

Documents

04-914

Sigma-Aldrich

Anti-Smad2/3 Antibody, clone C4T, rabbit monoclonal

culture supernatant, clone C4T, from rabbit

Synonym(s):

Deletion target in pancreatic carcinoma 4, MAD, mothers against decapentaplegic homolog 4, MAD, mothers against decapentaplegic homolog 4 (Drosophila), Mothers against DPP homolog 4, SMAD 4, SMAD family member 4, SMAD, mothers against DPP homolog 4, SMAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

C4T, monoclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SMAD2(4087)

General description

Smad proteins are regulators of transcription which transduce signals from TGFβ Receptors. Smad proteins homotrimerize, and when activated, two distinct homotrimers assemble into a heterosextamer. Smad proteins fall into three classes. The receptor-regulated Smad proteins, Smad 1, 2, 3, 5, and 9 couple to specific receptors and are phosphorylated by those receptors. Phosphorylated receptor-regulated Smad proteins then bind to a co-Smad, such as Smad4/DPC4, and the complex moves to the nucleus where it associates with FAST-1 to stimulate target gene transcription. A third class of Smad proteins is the inhibitory group of Smad 6, 7, 8. Smad proteins have Mad-homology domains 1 and 2 (MH1 and MH2). MH1 domains are involved in DNA binding, while MH2 domains function in homotrimerization, receptor interaction and phosphorylation. Smad 4 mutations are frequently found in cancer, all of which cluster to the MH1 and MH2 domains of the protein. Those in the MH2 domain affect the ability of the protein to homotrimerize. The phosphorylation of Smad proteins is the regulatory signal in their activation, and can be monitored by the use of phosphorylation state-specific antibodies.

Specificity

Predicted to cross-react with mouse and rat based on conservation of immunogen sequence.
Recognizes Smad2, Mr 55-60 kDa and Smad3, Mr 50 kDa.

Immunogen

Epitope: a.a. 186-273
Fusion protein corresponding to amino acids 186-273 of human Smad2.

Application

Research Category
Signaling
Research Sub Category
Transcription Factors
Use Anti-Smad2/3 Antibody, clone C4T (Rabbit Monoclonal Antibody) validated in WB to detect Smad2/3 also known as Deletion target in pancreatic carcinoma 4, mothers against decapentaplegic homolog 4.

Quality

Evaluated by Western Blot on L6 lysates.
Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Smad2/3 in L6 cell lysate.

Target description

~50-60 kDa

Linkage

Replaces: 05-914

Physical form

Cultured supernantant containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RIPA lysates from L6 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan-Hua Tsai et al.
Cells, 10(11) (2021-11-28)
Probiotics are defined as microorganisms with beneficial health effects when consumed by humans, being applied mainly to improve allergic or intestinal diseases. Due to the increasing resistance of pathogens to antibiotics, the abuse of antibiotics becomes inefficient in the skin
Catherine Koch et al.
Molecular therapy. Methods & clinical development, 17, 1178-1189 (2020-06-10)
Myotubular myopathy, also called X-linked centronuclear myopathy (XL-CNM), is a severe congenital disease targeted for therapeutic trials. To date, biomarkers to monitor disease progression and therapy efficacy are lacking. The Mtm1-/y mouse is a faithful model for XL-CNM, due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service