Skip to Content
Merck
All Photos(1)

Documents

T-073

Supelco

L-Thyroxine (T4) solution

100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

L-Thyroxine, 3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Beilstein:
2228515
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.1N NH3

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

223 °C (dec.) (lit.)

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1

InChI key

XUIIKFGFIJCVMT-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Thyroxine or T4 is a thyroid hormone responsible for regulation of the body′s basal metabolic rate and essential for growth, development and heart and nervous system function. Free and total T4 serum levels are measured by LC-MS/MS for determination of hyper- or hypothyroidism as well as for monitoring treatment with synthetic thyroid hormones. This Certified Spiking Solution® is suitable for use in LC-MS/MS testing methods as starting materials in preparation of linearity standards, calibrators, and controls.

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and, thus, protein synthesis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Sutherland et al.
The Journal of endocrinology, 65(3), 319-332 (1975-06-01)
A competitive binding technique is described for the estimation of the thyroxine (T4)-binding properties of serum proteins in dilute blood serum and lymph. When used in conjunction with an assay for total T4 the following parameters can be estimated: the
Ihor Hrytsiuk et al.
Archives of pediatrics & adolescent medicine, 156(5), 485-491 (2002-05-01)
To determine the effect of levothyroxine sodium starting dose on cognitive development, growth, or behavior in children with congenital hypothyroidism identified by neonatal screening. Systematic review of cohort studies. Two analyses were performed: a between-study comparison of mean starting dose
L Ashley Watson et al.
The Journal of clinical investigation, 123(5), 2049-2063 (2013-04-09)
Human ATRX mutations are associated with cognitive deficits, developmental abnormalities, and cancer. We show that the Atrx-null embryonic mouse brain accumulates replicative damage at telomeres and pericentromeric heterochromatin, which is exacerbated by loss of p53 and linked to ATM activation.
Sonja Heyerdahl et al.
Thyroid : official journal of the American Thyroid Association, 13(11), 1029-1038 (2003-12-04)
Neonatal screening programs for congenital hypothyroidism (CH) were initiated in the 1970s to ensure early treatment in order to prevent mental retardation. With screening, developmental prognosis is considerably improved, but follow-up studies still report developmental delay compared to controls. To
Aleid G van Wassenaer et al.
Clinical endocrinology, 56(5), 621-627 (2002-05-29)
Thyroid hormone is crucial for brain development during foetal and neonatal life. In very preterm infants, transient low levels of plasma T4 and T3 are commonly found, a phenomenon referred to as transient hypothyroxinaemia of prematurity. We investigated whether breast

Protocols

Determination of the thyroid hormones T4, T3 and rT3 from plasma using an Ascentis Express RP-Amide cartridge to trap the analytes, and a Phenyl-Hexyl column to resolve them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service