Skip to Content
Merck
All Photos(1)

Key Documents

878110C

Avanti

C16-02:0 PC

Avanti Research - A Croda Brand 878110C

Synonym(s):

1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54NO7P
CAS Number:
Molecular Weight:
523.68
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (878110C-5mg)
pkg of 1 × 2.5 mL (878110C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878110C

concentration

10 mg/mL (878110C-25mg)
5 mg/mL (878110C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C26H54NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5/h26H,6-24H2,1-5H3/t26-/m1/s1

InChI key

HVAUUPRFYPCOCA-AREMUKBSSA-N

General description

C16-02:0 PC, also known as 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine (C16 PAF) (acetyl PAF), is a platelet-activating factor (PAF). Acetyl PAF is present in the body and it contains ether-linked hexadecyl chain at the sn-1 position and an acetyl group at the sn-2 position.

Application

C16-02:0 PC or 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine, a platelet-activating factor (PAF) is used as a substrate for the determination of platelet-activating factor-acetylhydrolase (PAF-AH) activity in plasma or serum.

Biochem/physiol Actions

Platelet-activating factor (PAF) is a pro-inflammatory agent that exhibits variety of biological and pharmacological effects. It plays a major role in initiation and progression of atherosclerosis. PAF acts as a potential biomarker for coronary artery diseases. It yields LysoPAF (1-O-alkyl-2-hydroxy-sn-glycero-3-phosphocholine) on hydrolysis by platelet-activating factor-acetylhydrolase.

Packaging

5 mL Clear Glass Sealed Ampule (878110C-25mg)
5 mL Clear Glass Sealed Ampule (878110C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana M Stafforini et al.
Free radical biology & medicine, 59, 100-107 (2012-06-05)
This article presents a radiometric assay to determine the enzymatic activity of platelet-activating factor (PAF) acetylhydrolase (PAF-AH), also known as lipoprotein-associated phospholipase A2 and phospholipase A2 group 7A. The method is based on the release of radioactively labeled acetate from
Sunil A Nankar et al.
Biochimie, 95(6), 1196-1207 (2013-02-05)
Apolipoprotein-derived peptides are promising candidates for the treatment of various inflammatory conditions and the main mechanism proposed for the protective action of these peptides includes binding to pro-inflammatory lipid mediators with high affinity and facilitating their sequestration/metabolism/clearance in the body.
Saki Yamaura et al.
Clinica chimica acta; international journal of clinical chemistry, 481, 184-188 (2018-03-20)
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service