Skip to Content
Merck
All Photos(1)

Key Documents

870292P

Avanti

16:0 PA-PEG3-mannose

Avanti Research - A Croda Brand 870292P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phospho((ethyl-1′,2′,3′-triazole)triethyleneglycolmannose) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H99N4O16P
CAS Number:
Molecular Weight:
1055.32
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (870292P-1mg)
pkg of 1 × 10 mg (870292P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870292P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@H]1O[C@@H](OCCOCCOCCN(N=N2)C=C2CCOP(OC[C@H](OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)([O-])=O)[C@@H](O)[C@H](O)[C@H]1O.[NH4+]

General description

16:0 PA-PEG3-mannose, also known as 1,2-dipalmitoyl-sn-glycero-3-phospho((ethyl-1′,2′,3′-triazole)triethyleneglycolmannose) is a head group modified functionalized lipid.

Application

16:0 PA-PEG3-mannose or 1,2-dipalmitoyl-sn-glycero-3-phospho((ethyl-1′,2′,3′-triazole)triethyleneglycolmannose) has been used to link the mannose receptor on dendritic cells during the formation of lipopolyplexes (LPPs).

Packaging

5 mL Amber Glass Screw Cap Vial (870292P-10mg)
5 mL Amber Glass Screw Cap Vial (870292P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Perche et al.
Molecular therapy. Nucleic acids, 17, 767-775 (2019-08-26)
Nucleic acid vaccination relies on injecting DNA or RNA coding antigen(s) to induce a protective immune response. RNA vaccination is being increasingly used in preclinical and clinical studies. However, few delivery systems have been reported for in vivo delivery of RNA of different
David A Bryce et al.
Analytical chemistry, 90(19), 11509-11516 (2018-09-04)
Interactions of lectins, proteins that selectively bind carbohydrates, play an important role in many biological processes including cell adhesion, immune response, and cell signaling. Given the range of lectin functions and their potential for application in disease detection, there is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service