Skip to Content
Merck
All Photos(1)

Key Documents

W343307

Sigma-Aldrich

2-Methoxy-3-(1-methylpropyl)pyrazine

≥98%, FG

Synonym(s):

2-sec-Butyl-3-methoxypyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O
CAS Number:
Molecular Weight:
166.22
FEMA Number:
3433
EC Number:
Council of Europe no.:
11300
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.062
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.492 (lit.)

bp

50 °C/1 mmHg (lit.)

density

1 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; vegetable; pepper

SMILES string

CCC(C)c1nccnc1OC

InChI

1S/C9H14N2O/c1-4-7(2)8-9(12-3)11-6-5-10-8/h5-7H,4H2,1-3H3

InChI key

QMQDJVIJVPEQHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service