Recommended Products
biological source
Santalum album l.
Quality Level
grade
Halal
Kosher
natural
optical activity
[α]20/D −15°, neat
refractive index
n20/D 1.507 (lit.)
bp
276 °C (lit.)
density
0.974 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
woody; spicy; herbaceous; balsamic
Looking for similar products? Visit Product Comparison Guide
Preparation Note
Extraction method: steam distillation
Disclaimer
For R&D or non-EU Food use. Not for retail sale.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Repellent effect of santalol from sandalwood oil against Tetranychus urticae (Acari: Tetranychidae).
Journal of economic entomology, 105(2), 379-385 (2012-05-23)
Thirty-four essential oils were screened for their repellent activities against the twospotted spider mite, Tetranychus urticae Koch (Acarina: Tetranychidae), at 0.1% concentration level using choice and no-choice laboratory bioassays. Of these, 20 essential oils showed significant repellencies against T. urticae
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 421-432 (2007-11-06)
Sandalwood (Santalum album L.) is a fragrant wood from which oil is derived for use in food and cosmetics. Sandalwood oil is used in the food industry as a flavor ingredient with a daily consumption of 0.0074 mg/kg. Over 100
Lipids, 48(2), 105-113 (2013-01-01)
The aim of the present study was to investigate the effect of sandalwood seed oil on fatty acid (FA) profiles and inflammatory factors in rats. Fifty male Sprague-Dawley rats were randomly divided into five different dietary groups: 10 % soybean
Chemistry & biodiversity, 5(6), 1000-1010 (2008-07-12)
Natural sandalwood oil, a unique and valuable ingredient in fine perfumery, has been the focus of scientific interest for many years. Due to its scarcity and its high price, the search for novel synthetic raw materials imitating the characteristic odor
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 804-811 (2012-05-11)
The anticancer effects of α-santalol, a major component of sandalwood oil, have been reported against the development of certain cancers such as skin cancer both in vitro and in vivo. The primary objectives of the current study were to investigate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service