Skip to Content
Merck
All Photos(1)

Key Documents

N12007

Sigma-Aldrich

4-Nitrobenzonitrile

97%

Synonym(s):

1-Cyano-4-nitrobenzene, 4-Cyano-1-nitrobenzene, 4-Cyanonitrobenzene, p-Cyanonitrobenzene, p-Nitrobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CN
CAS Number:
Molecular Weight:
148.12
Beilstein:
972078
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

144-147 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(cc1)C#N

InChI

1S/C7H4N2O2/c8-5-6-1-3-7(4-2-6)9(10)11/h1-4H

InChI key

NKJIFDNZPGLLSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Maroz et al.
Organic & biomolecular chemistry, 8(2), 413-418 (2010-01-13)
The one-electron reduction chemistry of the antituberculosis drug PA-824, together with a series of closely related compounds, has been investigated in irradiated anaerobic propan-2-ol solution. The protic solvent, of low dielectric constant, was chosen to mimic the environment of a
N Dorostkar-Ahmadi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1375-1380 (2011-06-07)
Reaction of 4-nitrobenzonitrile oxide (2) which was generated in situ with acrylo nitrile (3), methyl methacrylate (4) and allyl bromide (5) as dipolarphile afforded the new 7a, 8a and 9a compounds respectively. Reactivity and regiochemistry of these reactions were investigated
G Monechi et al.
La Medicina del lavoro, 82(2), 137-141 (1991-03-01)
The paper describes a singular case of acute intoxication by 4-nitrobenzonitrile, a rare intermediate compound produced by pharmaceutical synthesis. The patients subsequently showed an increase in triglycerides and fatty alterations of the liver.
B Markus et al.
Journal of pharmaceutical sciences, 83(12), 1729-1734 (1994-12-01)
Studies on the metabolic fate of aromatic nitriles, in contrast to their aliphatic counterparts, have been minimal and the subject of controversy. The in vitro metabolic fate of several aromatic nitriles with varying substituents was investigated by using rat liver
A P Kumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 53A(12), 2041-2048 (1998-01-23)
A zero-order normal coordinate analysis of both the in-plane and out-of-plane vibrations was made for 2-chloro, 6-fluorobenzonitrile, s-trichlorobenzonitrile, p- and m-methoxybenzonitriles and m-nitrobenzonitrile, transferring the force constants from our earlier work. The observed and calculated frequencies agree with an average

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service