Skip to Content
Merck
All Photos(1)

Key Documents

M7707

Sigma-Aldrich

2-Mesitylenesulfonyl chloride

99%

Synonym(s):

2,4,6-Trimethylbenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2SO2Cl
CAS Number:
Molecular Weight:
218.70
Beilstein:
1107601
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

mp

55-57 °C (lit.)

SMILES string

Cc1cc(C)c(c(C)c1)S(Cl)(=O)=O

InChI

1S/C9H11ClO2S/c1-6-4-7(2)9(8(3)5-6)13(10,11)12/h4-5H,1-3H3

InChI key

PVJZBZSCGJAWNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Mesitylenesulfonyl chloride, also known as 2,4,6-Trimethylbenzenesulfonyl chloride, is an organic compound that is commonly used as a coupling agent in the nucleophilic substitution reaction to synthesize novel phospholipid derivatives of botulin and cyclodextrin.

Application

Coupling reagent in polynucleotide synthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A family of single-isomer, dicationic cyclodextrin chiral selectors for capillary electrophoresis: M ono-6A-ammonium-6C-butylimidazolium-beta-cyclodextrin chlorides
Yun D, et al.
Electrophoresis, 34, 833-840 (2013)
Tetrahedron, 48, 1729-1729 (1992)
Stéphane Menuel et al.
Beilstein journal of organic chemistry, 16, 2598-2606 (2020-11-03)
The mechanically assisted synthesis of organic compounds has recently focused considerable attention as it may be unique in features to selectively direct the reaction pathway. In the continuation of our work on the synthesis of modified cyclodextrins (CDs) via mechanochemical
Synthesis of 28-O-(1, 2-Diacyl-SN-glycero-3-phospho)-betulin
B Tubek, et al.
Synthetic Communications, 42, 3648-3654 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service