Skip to Content
Merck
All Photos(1)

Documents

M47949

Sigma-Aldrich

2-Methylheptane

98%

Synonym(s):

2-Methylheptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(CH3)2
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696862
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.77 psi ( 37.7 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

116 °C/761 mmHg (lit.)

density

0.698 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(C)C

InChI

1S/C8H18/c1-4-5-6-7-8(2)3/h8H,4-7H2,1-3H3

InChI key

JVSWJIKNEAIKJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel P Killeen et al.
Journal of agricultural and food chemistry, 65(17), 3551-3558 (2017-04-20)
Fish oils are the primary dietary source of ω-3 polyunsaturated fatty acids (PUFA), but these compounds are prone to oxidation, and commercial fish oil supplements sometimes contain less PUFA than claimed. These supplements are predominantly sold in softgel capsules. In
Feras Abujaber et al.
Talanta, 201, 266-270 (2019-05-28)
Magnetic cellulose nanoparticles (MCNPs) were used for the first time as sorbents for stir bar-sorptive dispersive microextraction (SBSDME). The main experimental parameters involved in the SBSDME process that affect extraction (i.e. amount of MCNPs, ionic strength, pH, extraction and back-extraction
Isabel Natalia Sierra-Garcia et al.
Extremophiles : life under extreme conditions, 21(1), 211-229 (2016-12-05)
Microorganisms have shown their ability to colonize extreme environments including deep subsurface petroleum reservoirs. Physicochemical parameters may vary greatly among petroleum reservoirs worldwide and so do the microbial communities inhabiting these different environments. The present work aimed at the characterization
Naser Mohammadpour Dounighi et al.
Current pharmaceutical design, 23(18), 2665-2672 (2017-01-13)
There is no doubt about the whole cell pertussis vaccine efficacy, but it is necessary to improve the vaccine quality specially to decrease its toxicity by obtaining good immunogenicity with low bacterial content. In this work, under optimum condition inactivated
Sultan Hassan Alamri et al.
International journal of environmental research and public health, 18(5) (2021-04-04)
To control the spread of coronavirus disease (COVID-19), Saudi Arabia's government imposed a strict lockdown during March-July 2020. As a result, the public was confined to indoors, and most of their daily activities were happening in their indoor places, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service