Skip to Content
Merck
All Photos(2)

Key Documents

GF93473118

Hafnium

foil, 25x25mm, thickness 0.5mm, annealed, 97%

Synonym(s):

Hafnium, HF000230

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
MDL number:
UNSPSC Code:
12141718
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

foil

manufacturer/tradename

Goodfellow 934-731-18

resistivity

29.6 μΩ-cm, 0°C

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service