GF37996789
Gold
tube, 200mm, outside diameter 2.0mm, inside diameter 1.7mm, wall thickness 0.15mm, as drawn, 99.95%
Synonym(s):
Gold, AU007201
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99.95%
form
tubes
manufacturer/tradename
Goodfellow 379-967-89
resistivity
2.05 μΩ-cm, 0°C
L × wall thickness
200 mm × 0.15 mm
O.D. × I.D.
2.0 mm × 1.7 mm
bp
2808 °C (lit.)
mp
1063 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[Au]
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Journal of nanoscience and nanotechnology, 14(7), 4851-4857 (2014-04-25)
The current study involves in identification and molecular levels characterization of optimal size and concentration of gold nanoparticles (AuNPs). Stable, gold nanoparticles were synthesized and characterized using transmission electron microscopy (TEM) and dynamic light scattering (DLS). The concentration and size
Journal of nanoscience and nanotechnology, 14(6), 4029-4035 (2014-04-18)
Water soluble BSA-stabilized gold nanoclusters (Au NCs) were synthesized with a simple one-pot procedure. The as-prepared Au NCs were able to emit intensive red fluorescence under the excitation of ultraviolet light, and the fluorescence could be quenched by enzymatic hydrolysis.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Journal of nanoscience and nanotechnology, 14(6), 4078-4081 (2014-04-18)
Colloidal gold is extensively used for molecular sensing because that the surface plasmon resonance (SPR) bands are affected by changes in the dielectric properties in the close vicinity of these structures due to the binding of ligands to the corresponding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service