Skip to Content
Merck
All Photos(2)

Key Documents

GF35961655

Cerium

wire reel, 100mm, diameter 1.0mm, hard, 99.9%

Synonym(s):

Cerium, CE005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
MDL number:
UNSPSC Code:
12141601
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 359-616-55

resistivity

73 μΩ-cm, 20°C

L × diam.

100 mm × 1.0 mm

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flemming R Cassee et al.
Critical reviews in toxicology, 41(3), 213-229 (2011-01-20)
Advances of nanoscale science have produced nanomaterials with unique physical and chemical properties at commercial levels which are now incorporated into over 1000 products. Nanoscale cerium (di) oxide (CeO(2)) has recently gained a wide range of applications which includes coatings
Soumen Das et al.
Nanomedicine (London, England), 8(9), 1483-1508 (2013-08-31)
Promising results have been obtained using cerium (Ce) oxide nanoparticles (CNPs) as antioxidants in biological systems. CNPs have unique regenerative properties owing to their low reduction potential and the coexistence of both Ce(3+)/Ce(4+) on their surfaces. Defects in the crystal
C J Van Noorden et al.
Journal of microscopy, 171(Pt 1), 3-16 (1993-07-01)
Methods based on the use of cerium to detect the activity of oxidases and phosphatases are rapidly expanding. Both classes of enzymes can be demonstrated with cerium at the electron and light microscopical level. The in situ detection of H2O2
Ilse Gosens et al.
Nanotoxicology, 8(6), 643-653 (2013-06-19)
There are many uncertainties regarding the hazard of nanosized particles compared to the bulk material of the parent chemical. Here, the authors assess the comparative hazard of two nanoscale (NM-211 and NM-212) and one microscale (NM-213) cerium oxide materials in
Srinivas Aalapati et al.
Nanotoxicology, 8(7), 786-798 (2013-08-07)
Male CD1 mice were subjected to nose-inhalation exposure of CeO2 nanoparticles (NPs) for 0, 7, 14 or 28 days with 14 or 28 days of recovery time at an aerosol concentration of 2 mg/m(3). Markers of lung injury and pro-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service