Documents
ESYNTH004
Sigma-Aldrich
SynLectro 50 mL Jacketed Glass Cell with PTFE stopper
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
General description
SynLectro 50 mL Jacketed Glass Cell with PTFE Stopper is the part of the SynLectro Electrolysis Platform for synthetic electrochemistry.
Features and Benefits:
  • Designed to simplify setup, optimize reactivity, and improve safety
  • Side-arm enables connection to reflux condenser or inert gas
  • Jacket supports thermal control
  • Chemical resistant material (PTFE)
  • Protects electrical contacts from electrolytes
  • Sleeve (optional) prevents seized joints
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Related Content
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.