Skip to Content
Merck
All Photos(2)

Documents

B102628

Sigma-Aldrich

4-tert-Butyltoluene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CH3
CAS Number:
Molecular Weight:
148.24
Beilstein:
2038670
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

191 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)C(C)(C)C

InChI

1S/C11H16/c1-9-5-7-10(8-6-9)11(2,3)4/h5-8H,1-4H3

InChI key

QCWXDVFBZVHKLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ingebrigtsen et al.
Acta pharmacologica et toxicologica, 51(3), 203-208 (1982-09-01)
The distribution of [methyl-14C]p-tert.-butyltoluene was studied in the rat by use of whole-body autoradiography and liquid scintillation counting. The test substance was administered by inhalation. High concentration of radioactivity was seen in the CNS immediately after inhalation, from where it
S P Lund et al.
International journal of psychophysiology : official journal of the International Organization of Psychophysiology, 14(1), 41-48 (1993-01-01)
The use of evoked potentials to measure neurotoxicity was evaluated using 4-tert-butyltoluene (TBT) as a test compound. Male Wistar rats were habituated to the recordings of auditory- and flash-evoked potentials until the combined waveform of the evoked potentials reached a
H R Lam et al.
Pharmacology & toxicology, 87(1), 11-17 (2000-09-15)
Long-lasting central nervous system (CNS) neurotoxicity of 4-tert-butyltoluene (TBT) has been investigated using electrophysiology, behaviour, and neurochemistry in Long Evans rats exposed by inhalation to 0, 20, or 40 p.p.m. TBT 6 hr/day, 7 days/week for 4 weeks. Flash evoked
Uptake, distribution and elimination of p-tert-butyltoluene (TBT) in mice by inhalation.
A Rasmussen et al.
Acta pharmacologica et toxicologica, 47(3), 236-238 (1980-09-01)
A Walde et al.
Acta pharmacologica et toxicologica, 53(1), 57-63 (1983-07-01)
The metabolism of p-tert.-butyltoluene (TBT) was studied in the rat and guinea pig. Both the methyl and the tert.-butyl group were oxidized to alcohol and carboxylic acid derivatives in these species. The major urinary metabolites in rats were p-tert.-butylbenzoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service