Skip to Content
Merck
All Photos(1)

Documents

A83008

Sigma-Aldrich

3,4-Dimethoxyaniline

98%

Synonym(s):

4-Aminopyrocatechol dimethyl ether, 4-Aminoveratrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(OCH3)2
CAS Number:
Molecular Weight:
153.18
Beilstein:
743399
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

174-176 °C/22 mmHg (lit.)

mp

85-89 °C (lit.)

SMILES string

COc1ccc(N)cc1OC

InChI

1S/C8H11NO2/c1-10-7-4-3-6(9)5-8(7)11-2/h3-5H,9H2,1-2H3

InChI key

LGDHZCLREKIGKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Ikawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(19), 4320-4332 (2020-01-03)
Benzynes were selectively generated in situ from phenols and trapped regioselectively with potassium hexamethyldisilazide to form primary anilines following acidic workup. The direct conversion of a phenolic hydroxyl group into a free amino group is a useful method for the
Daniela Iguchi et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 15(1), 105-116 (2015-12-24)
Photochemical reactions tend to give more than one photoproduct. However, such a reaction can be a powerful synthetic tool when it is possible to conduct it in regioselective conditions yielding a single photoproduct. Water-surfactant solutions as reaction media can be
Chad D Iverson et al.
Journal of chromatography. A, 1422, 186-193 (2015-10-28)
Porous graphitic carbon (PGC) is an increasingly popular and attractive phase for HPLC on account of its chemical and thermal stability, and its unique separation mechanism. However, native PGC is strongly hydrophobic and in some instances excessively retentive. As part

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service