Skip to Content
Merck
All Photos(1)

Key Documents

930067

Sigma-Aldrich

2-(Pyrimidin-5-yl)benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N2O
Molecular Weight:
185.20
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.06

form

powder or chunks

Quality Level

mp

119-120 °C

storage temp.

−20°C

Application

2-(Pyrimidin-5-yl)benzaldehyde is a temporary directing group (TDG) to assist as a co-catalyst for metal catalyzed C-H functionalization. Often in C-H functionalization, an auxiliary compound is used to control site selectivity. These traditionally are covalently bonded to the compound of interest, and must subsequently be removed after functionalization, like a typical protecting group. To simplify the process of C-H functionalization, 2-fluoro-6-(pyrimidin-5-yl)aniline is one of a series of temporary directing groups developed by Deb Maiti′s lab that promote site selectivity without the inclusion of additional synthetic steps.


2-(pyrimidin-5-yl)benzaldehyde is an effective TDG for meta directed C-H functionalization of amine substituted target compounds, with high selectivity.

Other Notes

Imine as a linchpin approach for meta-C–H functionalization

https://www.nature.com/articles/s41570-021-00311-3">Transient directing ligands for selective metal-catalysed C–H activation

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukdev Bag et al.
Nature communications, 12(1), 1393-1393 (2021-03-04)
Despite the widespread applications of C-H functionalization, controlling site selectivity remains a significant challenge. Covalently attached directing groups (DGs) served as ancillary ligands to ensure ortho-, meta- and para-C-H functionalization over the last two decades. These covalently linked DGs necessitate
Transient directing ligands for selective metal-catalysed C?H activation
Nupur, et al.
Nature Reviews Chemistry, 5, 646?659-646?659 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service