926590
Upconversion Nanoparticles
NaYF4-Yb,Tm@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980, blue light
Synonym(s):
UCNPs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
Quality Level
concentration
2 mg/mL in water
size
30 nm ± 5 nm
matrix active group
PEG-COOH surface treatment
fluorescence
λex 980 (blue)
λex 980 nm; λem 360 nm
λem 450 nm
λem 470 nm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Upconversion nanoparticles (ucnps), are the result of a unique optical process in which near-infrared (NIR) light excitation is converted into visible and/or UV emission. Upconversion (UC) utilizes sequential absorption of multiple photons utilizing both lifetime and real ladder-like energy levels of ions in a host lattice to produce a higher energy anti-Stokes luminescence.
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
Applications include:
Fluorescent microscopy
Deep-tissue bioimaging
Nanomedicine
Optogenetics
Security labelling
Volumetric display
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
Applications include:
Fluorescent microscopy
Deep-tissue bioimaging
Nanomedicine
Optogenetics
Security labelling
Volumetric display
Legal Information
Product of RuixiBiotechCo. Ltd
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Advances in highly doped upconversion nanoparticles
Wen, S., Zhou, J., Zheng, K. et al.
Nature Communications, 9, 2415-2415 (2018)
Engineered upconversion nanoparticles for resolving protein interactions inside living cells
Drees, C. et al.
Angewandte Chemie (International Edition in English), 55, 11668?11672-11668?11672 (2016)
Upconversion nanoparticles: design, nanochemistry, and applications in theranostics
Chen G., et al.
Chemical Reviews, 114, 5161-5214 (2014)
Amplified stimulated emission in upconversion nanoparticles for super-resolution nanoscopy
Liu, Y. et al.
Nature, 543, 229?233-229?233 (2017)
Engineering bright sub-10-nm upconverting nanocrystals for single-molecule imaging
Gargas, D. J. et al.
Nature Nanotechnology, 9, 300?305-300?305 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service