Skip to Content
Merck
All Photos(2)

Documents

913006

Sigma-Aldrich

IEICO

≥99%

Synonym(s):

2,2′-((2Z,2′Z)-((5,5′-bis(4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b′]dithiophene-2,7-diyl)bis(4-((2- ethylhexyl)oxy)thiophene-5,2-diyl))bis(methanylylidene))bis-(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile, 2,2′-[[4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b′]dithiophene-2,7-diyl]bis[[4-[(2-ethylhexyl)oxy]-5,2-thiophenediyl]-(Z)-methylidyne(3-oxo-1H-indene-2,1(3H)-diylidene)]]bis-propanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C114H118N4O4S4
CAS Number:
Molecular Weight:
1736.44
UNSPSC Code:
12352101
NACRES:
NA.23

description

Band gap: Eg=1.34 eV

Quality Level

Assay

≥99%

form

solid

color

dark

Orbital energy

HOMO -5.32 eV 
LUMO -3.95 eV 

Application

IEICO is a high-performance narrow-bandgap non-fullerene acceptor (NFA) with a complementary absorption spectrum to many of the best medium to high bandgap OPV devices. This feature was used to create high-efficiency multi-layer tandem solar cells, reaching a PCE of nearly 14%. Over 11% PCE can be reached in single junction cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunpeng Qin et al.
Advanced materials (Deerfield Beach, Fla.), 29(24) (2017-05-04)
Tandem organic solar cells (TOSCs), which integrate multiple organic photovoltaic layers with complementary absorption in series, have been proved to be a strong contender in organic photovoltaic depending on their advantages in harvesting a greater part of the solar spectrum
Huifeng Yao et al.
Advanced materials (Deerfield Beach, Fla.), 28(37), 8283-8287 (2016-07-06)
A novel non-fullerene acceptor, possessing a very low bandgap of 1.34 eV and a high-lying lowest unoccupied molecular orbital level of -3.95 eV, is designed and synthesized by introducing electron-donating alkoxy groups to the backbone of a conjugated small molecule.
Runnan Yu et al.
Advanced materials (Deerfield Beach, Fla.), 29(26) (2017-05-04)
High-performance ternary organic solar cells are fabricated by using a wide-bandgap polymer donor (bithienyl-benzodithiophene-alt-fluorobenzotriazole copolymer, J52) and two well-miscible nonfullerene acceptors, methyl-modified nonfullerene acceptor (IT-M) and 2,2'-((2Z,2'Z)-((5,5'-(4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydros-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene-5,2-diyl))bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile (IEICO). The two acceptors with complementary absorption spectra and similar lowest unoccupied molecular
Yong Cui et al.
Journal of the American Chemical Society, 139(21), 7302-7309 (2017-05-13)
Fabricating organic solar cells (OSCs) with a tandem structure has been considered an effective method to overcome the limited light absorption spectra of organic photovoltaic materials. Currently, the most efficient tandem OSCs are fabricated by adopting fullerene derivatives as acceptors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service