Skip to Content
Merck
All Photos(1)

Key Documents

909130

Sigma-Aldrich

Sulfonated reduced graphene oxide

sodium salt

Synonym(s):

Dispersibe rGO, Dispersible reduced graphene oxide, rGO sulfate, rGO-SO3Na

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352119
NACRES:
NA.23

form

powder

Application

Sulfonated reduced graphene oxide (sodium salt) is a specially designed highly dispersible form of reduced graphene oxide (rGO) salt. This product offers conductive rGO sheets with abundant sulfate groups, making it a versatile starting material for various applications:
  • Sulfonated graphene was reported to show potential as high performance energy materials for high-energy-density Li-S batteries.
  • It is a promising solid-phase acid catalyst that can be recycled after reaction and still maintain its catalytical properties for various chemical reactions including the production of biodiesel.
  • Different types of high performance sensors have been fabricated using sulfonated reduced graphene oxide.
  • The dispersity and the reactivity of the sulfate groups further allow feasible synthesis of crosslinkable functional composite.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

δ-MnO2 nanoflowers on sulfonated graphene sheets for stable oxygen reduction and hydrogen evolution reaction
Begum H, et al.
Electrochimica Acta, 296, 235-242 (2019)
Enhanced hydrolysis of cellulose by highly dispersed sulfonated graphene oxide.
Huang L, et al.
BioResources, 13(4), 8853-8870 (2018)
Ruma Perveen et al.
Scientific reports, 7(1), 13353-13353 (2017-10-19)
The proposed work intended to make an intellectual contribution to the domain of green nanotechnology which emphasizes the chemical synthesis of a conducting nanocomposite based on the incorporation of gold nanoparticles (Au) into the redox matrix of polyindole (PIn) along
Synthesis of sulfonated graphene/carbon nanotubes/manganese dioxide composite with high electrochemical properties.
Li W, et al.
Ionics, 25, 999-1006 (2018)
Biodiesel production from palm oil using sulfonated graphene catalyst.
Nongbe M, et al.
Renewable Energy, 106135-141, 135-141 (2017)

Articles

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service