Skip to Content
Merck
All Photos(1)

Key Documents

901768

Sigma-Aldrich

Boron nitride suspension

Synonym(s):

2D-BN, 2D-Boron nitride, 2D-Boron nitride dispersion with nonionic surfactant, 2D-hBN, Hexagonal boron nitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BN
UNSPSC Code:
12352302
NACRES:
NA.23

description

100-2000 nm
thickness < 3 layers

form

liquid

concentration

1 mg/mL in H2O

Looking for similar products? Visit Product Comparison Guide

General description

Boron nitride suspension (h-BN) (1 mg/mL in water) has a hexagonal bonded structure, with high mechanical strength and good thermal conductivity. It belongs to the class of hexagonal layered materials. It has a direct band gap of 5.8 eV and can be used as a dielectric layer in electronic devices.

Application

2D-hexagonal boron nitride (2D-hBN) is a structural isomorph of graphene and it possesses high chemical, mechanical and thermal stability. However, unlike graphene, the 2D-hBN is a high band gap material. The 2D-hBN exhibits exotic optical and electrical properties and find applications in field effect transistors (FETs), photoelectric devices and UV detectors.
h-BN can be used in the fabrication of photonic devices. It can also be used as a multifunctional additive in a polymeric electrolyte for the formation of lithium metal batteries.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

2D boron nitride nanoflakes as a multifunctional additive in gel polymer electrolytes for safe, long cycle life and high rate lithium metal batteries.
Shim J, et al.
Energy & Environmental Science, 10(9), 1911-1916 (2017)
Boron nitride substrates for high-quality graphene electronics.
Dean CR, et al.
Nature Nanotechnology, 5(10), 722-722 (2010)
Hexagonal boron nitride for deep ultraviolet photonic devices.
Jiang HX and Lin JY
Semiconductor Science and Technology, 29(8), 084003-084003 (2014)
Two dimensional hexagonal boron nitride (2D-hBN): Synthesis, properties and applications.
Zhang K, et al.
Journal of Material Chemistry C, 5, 11992-11992 (2017)
The two-dimensional phase of boron nitride: Few-atomic-layer sheets and suspended membranes.
Pacile D, et al.
Applied Physics Letters, 92(13), 133107-133107 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service