Skip to Content
Merck
All Photos(1)

Documents

900448

Sigma-Aldrich

Graphene dispersion

greener alternative

≥0.2 mg/mLin DMF, sheet resistance 2 kΩ/sq, avg. no. of layers, 1 ‑ 3

Synonym(s):

Conductive ink, Electrochemically exfoliated graphene ink, Graphene ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

product name

Graphene dispersion, ≥0.2 mg/mL in DMF, sheet resistance 2 kΩ/sq, avg. no. of layers, 1 ‑ 3

description

Dispersion in DMF

Quality Level

form

dispersion

feature

avg. no. of layers 1 ‑ 3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥0.2 mg/mL (electrochemically exfoliated graphene)
≥0.2 mg/mL in DMF

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

  • Graphene preparation method: electrochemical exfoliation.
  • Graphene thickness by AFM: ∼80% 1-3 layers.
  • Sheet size by AFM: 5-10 μm.
  • Oxygen content: 5.5 at.% (C/O-ratio: 17.2).
  • Raman I_D/I_G ratio: 0.25.
  • Sheet resistance: 2 kΩ/sq.
  • Typical properties of films produced thereof (after dilution to 0.1 mg/mL and dry transfer from PTFE-filter):
6 nm film: 91% transmittance, sheet resistance of 24.2 kΩ/sq (as made), 7.9 kΩ/sq (after 30 min, 200 °C), <1 kΩ/sq (conc. HNO3-treatment).
16 nm film: 73% transmittance, sheet resistance of 7.6 kΩ/sq (as made), 1.8 kΩ/sq (after 30 min, 200 °C), <500 Ω/sq (conc. HNO3-treatment).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Graphene inks are highly conductive, low cost and super flexible. Click here for more information.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

131.0 °F

Flash Point(C)

55 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khaled Parvez et al.
Journal of the American Chemical Society, 136(16), 6083-6091 (2014-04-02)
Mass production of high-quality graphene sheets is essential for their practical application in electronics, optoelectronics, composite materials, and energy-storage devices. Here we report a prompt electrochemical exfoliation of graphene sheets into aqueous solutions of different inorganic salts ((NH4)2SO4, Na2SO4, K2SO4

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service