Skip to Content
Merck
All Photos(1)

Documents

86631

Sigma-Aldrich

Tetraethylammonium hydroxide solution

~25% in methanol (~1.5 M)

Synonym(s):

TEA hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(OH)
CAS Number:
Molecular Weight:
147.26
Beilstein:
3912961
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

~25% in methanol (~1.5 M)

anion traces

sulfate (SO42-): ≤1000 mg/kg

SMILES string

[OH-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1

InChI key

LRGJRHZIDJQFCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylammonium hydroxide (TEAOH) is a quaternary ammonium salt that is commonly used as a strong base and phase-transfer catalyst in organic synthesis.

Application

Tetraethylammonium hydroxide (25% in methanol) can be used as a base in the synthesis of 2-hydroxyacetophenoxime from 2-hydroxysalicylaldoxime and hydroxylamine hydrochloride. It is also used as an organic structure-directing agent in the synthesis of zeolites such as Beta, ZSM-20, and SAPO-34.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayabalan Nirmal et al.
Experimental eye research, 116, 27-35 (2013-07-31)
The present study was conducted to test the hypothesis; OCT may be active from blood-to-vitreous for the uptake of its substrates. Ocular uptake of Tetraethylammonium (TEA) across blood ocular barriers and the tissue distribution was evaluated in vivo in New
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Lei Yang et al.
The Journal of general physiology, 140(5), 529-540 (2012-10-31)
We investigated the effects of changing extracellular K(+) concentrations on block of the weak inward-rectifier K(+) channel Kir1.1b (ROMK2) by the three intracellular cations Mg(2+), Na(+), and TEA(+). Single-channel currents were monitored in inside-out patches made from Xenopus laevis oocytes
Ling Xiao et al.
Circulation research, 112(10), 1310-1322 (2013-03-28)
A chromosomal haplotype producing cardiac overexpression of dipeptidyl peptidase-like protein-6 (DPP6) causes familial idiopathic ventricular fibrillation. The molecular basis of transient outward current (I(to)) in Purkinje fibers (PFs) is poorly understood. We hypothesized that DPP6 contributes to PF I(to) and
Samuel J Goodchild et al.
The Journal of general physiology, 140(5), 495-511 (2012-10-17)
The open state of voltage-gated potassium (Kv) channels is associated with an increased stability relative to the pre-open closed states and is reflected by a slowing of OFF gating currents after channel opening. The basis for this stabilization is usually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service