Skip to Content
Merck
All Photos(2)

Documents

850187

Sigma-Aldrich

5-Bromouridine

98%

Synonym(s):

5-Bromouracil-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O6
CAS Number:
Molecular Weight:
323.10
Beilstein:
33664
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

optical activity

[α]22/D −11°, c = 2 in H2O

mp

180-182 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(Br)C(=O)NC2=O

InChI

1S/C9H11BrN2O6/c10-3-1-12(9(17)11-7(3)16)8-6(15)5(14)4(2-13)18-8/h1,4-6,8,13-15H,2H2,(H,11,16,17)/t4-,5-,6-,8-/m1/s1

InChI key

AGFIRQJZCNVMCW-UAKXSSHOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayu Hasegawa et al.
Mutation research, 699(1-2), 62-66 (2010-04-17)
Individuals with inherited xeroderma pigmentosum (XP) disorder and Cockayne syndrome (CS) are deficient in nucleotide excision repair and experience hypersensitivity to sunlight. Although there are several diagnostic assays for these disorders, the recovery of RNA synthesis (RRS) assay that can
Maria D Vibranovski et al.
Genetics, 186(1), 431-433 (2010-07-09)
Extensive gene expression during meiosis is a hallmark of spermatogenesis. Although it was generally accepted that RNA transcription ceases during meiosis, recent observations suggest that some transcription occurs in postmeiosis. To further resolve this issue, we provide direct evidence for
Hidenori Tani et al.
Genome research, 22(5), 947-956 (2012-03-01)
Mammalian genomes produce huge numbers of noncoding RNAs (ncRNAs). However, the functions of most ncRNAs are unclear, and novel techniques that can distinguish functional ncRNAs are needed. Studies of mRNAs have revealed that the half-life of each mRNA is closely
Monica Teta et al.
Diabetes, 54(9), 2557-2567 (2005-08-27)
Although many signaling pathways have been shown to promote beta-cell growth, surprisingly little is known about the normal life cycle of preexisting beta-cells or the signaling pathways required for beta-cell survival. Adult beta-cells have been speculated to have a finite
Anne Wilson et al.
Cell, 135(6), 1118-1129 (2008-12-09)
Bone marrow hematopoietic stem cells (HSCs) are crucial to maintain lifelong production of all blood cells. Although HSCs divide infrequently, it is thought that the entire HSC pool turns over every few weeks, suggesting that HSCs regularly enter and exit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service