Skip to Content
Merck
All Photos(1)

Key Documents

793671

Sigma-Aldrich

Ethylene-13C oxide

≥99 atom % 13C, ≥98% (CP), contains hydroquinone as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CCH4O
CAS Number:
Molecular Weight:
45.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

gas

contains

hydroquinone as stabilizer

mass shift

M+1

storage temp.

room temp

SMILES string

[13CH2]1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2/i1+1

InChI key

IAYPIBMASNFSPL-OUBTZVSYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service