761591
(4-(1,2,4,5-Tetrazin-3-yl)phenyl)methanamine hydrochloride
95%
Synonym(s):
Benzylamino tetrazine hydrochloride, H-Tz-Bz-NH3Cl hydrochloride
About This Item
Recommended Products
Assay
95%
form
solid
reaction suitability
reaction type: click chemistry
reagent type: linker
mp
206-225 °C (decomposition)
functional group
amine
storage temp.
−20°C
SMILES string
NCC1=CC=C(C2=NN=CN=N2)C=C1.[H]Cl
InChI
1S/C9H9N5.ClH/c10-5-7-1-3-8(4-2-7)9-13-11-6-12-14-9;/h1-4,6H,5,10H2;1H
InChI key
FAAUXGMWUKVOPI-UHFFFAOYSA-N
General description
Application
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service