Skip to Content
Merck
All Photos(1)

Key Documents

761133

Sigma-Aldrich

N,N-Diethyl-2-cyanoacetoamide

97%

Synonym(s):

2-Cyano-N,N-diethylacetamide, N,N-Diethylcyanoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O
CAS Number:
Molecular Weight:
140.18
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.466

density

1.013 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCN(CC)C(=O)CC#N

InChI

1S/C7H12N2O/c1-3-9(4-2)7(10)5-6-8/h3-5H2,1-2H3

InChI key

RYSHIRFTLKZVIH-UHFFFAOYSA-N

Application

N,N-Diethyl-2-cyanoacetoamide can be used:
  • To prepare N,N-diethyl 2-cyano-2-fluoro-2-phenylthioacetoamide, which is further used in the synthesis of various cyanofluoroamides.
  • In the synthesis of entacapone, a catechol-O-methyltransferase inhibitor by reacting with 3,4-dihydroxy-5-nitrobenzaldehyde.
  • As a building block in the synthesis of fluorophores, applicable as endoplasmic reticulum probes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-line reaction monitoring of entacapone synthesis by Raman spectroscopy and multivariate analysis
Novak P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 54(4), 660-666 (2011)
Manikowski, Andrzej et al.
Synthetic Communications, 39, 3621-3638 (2009)
A light-up endoplasmic reticulum probe based on a rational design of red-emissive fluorogens with aggregation-induced emission
Zhang C, et al.
Chemical Communications (Cambridge, England), 53(78), 10792-10795 (2017)
Bhawal, Baburao M., et al.
Synthetic Communications, 20, 3232-3243 (1990)
The development of a novel synthetic method for cyanofluoroamides using a cobaltadithiolene complex
Yokoyama Y, et al.
Synthesis, 2004(05), 701-705 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service