Skip to Content
Merck
All Photos(2)

Documents

723037

Sigma-Aldrich

2-Cyano-2-propyl dodecyl trithiocarbonate

97% (HPLC)

Synonym(s):

S-(2-Cyanoprop-2-yl)-S-dodecyltrithiocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31NS3
CAS Number:
Molecular Weight:
345.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

liquid

refractive index

n20/D 1.535

density

0.991 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCSC(=S)SC(C)(C)C#N

InChI

1S/C17H31NS3/c1-4-5-6-7-8-9-10-11-12-13-14-20-16(19)21-17(2,3)15-18/h4-14H2,1-3H3

InChI key

QSVOWVXHKOQYIP-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

2-Cyano-2-propyl dodecyl trithiocarbonate is used as a RAFT agent for controlled radical polymerization; especially suited for the polymerization of methacrylate, methacrylamide, and styrene monomers. It is also used as a Chain Transfer Agent (CTA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matías Regiart et al.
Analytica chimica acta, 963, 83-92 (2017-03-25)
We report a hybrid glass-poly (dimethylsiloxane) microfluidic immunosensor for epidermal growth factor receptor (EGFR) determination, based on the covalent immobilization of anti-EGFR antibody (anti-EGFR) on amino-functionalized mesoporous silica (AMS) retained in the central channel of a microfluidic device. The synthetized
Jeonghun Lee et al.
Journal of controlled release : official journal of the Controlled Release Society, 304, 164-172 (2019-05-15)
A blood clot (thrombus) is formed as a final product of the hemostatic process with two major components, a mesh of cross-linked fibrin and platelets activated by high concentration of hydrogen peroxide (H2O2). Thrombus formation impedes blood flow to brain
Hsiu-Pen Lin et al.
Macromolecular bioscience, 20(6), e2000049-e2000049 (2020-04-08)
Cationic polymers exhibit high cytotoxicity via strong interaction with cell membranes. To reduce cell membrane damage, a hydrophilic polymer is introduced to the cationic nanoparticle surface. The hydrophilic polymer coating of cationic nanoparticles resulted in a nearly neutral nanoparticle. These
Liqin Mei et al.
Journal of materials chemistry. B, 8(25), 5434-5440 (2020-06-13)
Zwitterionic polymers have attracted increasing attention due to their excellent fouling resistance ability and eco-friendliness. Yet, their non-degradability and hydrophilic nature limit their applications. In this study, we have prepared a novel surface-fragmenting hyperbranched copolymer with tertiary carboxybetaine ester (TCB)
Joseph L Mann et al.
Science translational medicine, 12(550) (2020-07-03)
Insulin has been used to treat diabetes for almost 100 years; yet, current rapid-acting insulin formulations do not have sufficiently fast pharmacokinetics to maintain tight glycemic control at mealtimes. Dissociation of the insulin hexamer, the primary association state of insulin

Articles

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

RAFT polymerization uses commercial agents to control polymer properties without cytotoxic heavy metals like ATRP.

See All

Protocols

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service