Skip to Content
Merck
All Photos(2)

Key Documents

721077

Sigma-Aldrich

Zinc oxide, dispersion

nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O

Synonym(s):

Zinc monoxide, Zinc white

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

20 wt. % in H2O

particle size

<100 nm (TEM)

avg. part. size

≤40 nm (APS)

pH

7.5±1.5

density

1.7 g/mL±0.1 g/mL at 25 °C

SMILES string

[Zn]=O

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zinc oxide, dispersion (ZnO) is a multi-function metal oxide with unique physio-chemical properties, which includes high chemical stability, broad range absorption spectra and high electrochemical characteristics. It can also be categorized as a semiconductor due to its high energy band width and high thermal stability that make it potentially useful in electronics and optoelectronics applications.

Application

ZnO dispersions have piezo and pyroelectric properties that make it useful in the development of sensors, generators, and photo-catalysis. It has low toxicity and biocompatibility, which facilitate its applications in biomedicine.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zinc oxide nanoparticles: chemical mechanisms and classical and non-classical crystallization
Ludi B and Niederberger M
Dalton Transactions, 42(35), 12554-12568 (2013)
A comprehensive review of ZnO materials and devices
Ozgur U, et al.
Journal of Applied Physics, 98(4), 11-11 (2005)
Splendid one-dimensional nanostructures of zinc oxide: a new nanomaterial family for nanotechnology
Wang ZL
ACS Nano, 2(10), 1987-1992 (2008)
Dispersing hydrophilic nanoparticles in hydrophobic polymers: HDPE/ZnO nanocomposites by a novel template-based approach.
de Luna MS, et al.
Express Polymer Letters, 8(5) (2014)
Electrical conduction and dielectric studies of ZnO pellets
Chaari M and Matoussi A
Physica B: Condensed Matter, 407(17), 3441-3447 (2012)

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service