Skip to Content
Merck
All Photos(2)

Key Documents

694509

Sigma-Aldrich

3,3-Difluoropyrrolidine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7F2N · HCl
CAS Number:
Molecular Weight:
143.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

133-136 °C

SMILES string

Cl.FC1(F)CCNC1

InChI

1S/C4H7F2N.ClH/c5-4(6)1-2-7-3-4;/h7H,1-3H2;1H

InChI key

YYVPZQADFREIFR-UHFFFAOYSA-N

Application

3,3-Difluoropyrrolidine hydrochloride can be used as a building block in the synthesis of:
  • Triazole substituted prolyl difluoropyrrolidines as potential inhibitors of dipeptidyl peptidase-4.
  • Dual leucine zipper kinase (DLK) inhibitors.

It can be also used as a reactant in the preparation of cyclic and acyclic β-aminofluoroalkenes via allylic amination using the Pd catalyst.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, Synthesis, Structure-Activity Relationships, and Docking Studies of 1-(?-1, 2, 3-Triazol Substituted Prolyl)-(S)-3, 3-Difluoropyrrolidines as a Novel Series of Potent and Selective Dipeptidyl Peptidase-4 Inhibitors
Zhang L, et al.
Chemical Biology & Drug Design, 81(2), 198-207 (2013)
Activation of Allylic C-F bonds: Palladium-Catalyzed Allylic Amination of 3, 3-Difluoropropenes
Pigeon X, et al.
Angewandte Chemie (International Edition in English), 49(6), 1123-1127 (2010)
Snahel Patel et al.
Journal of medicinal chemistry, 58(1), 401-418 (2014-10-24)
Dual leucine zipper kinase (DLK, MAP3K12) was recently identified as an essential regulator of neuronal degeneration in multiple contexts. Here we describe the generation of potent and selective DLK inhibitors starting from a high-throughput screening hit. Using proposed hinge-binding interactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service