Skip to Content
Merck
All Photos(1)

Key Documents

690481

Sigma-Aldrich

N-[3,5-Bis(trifluoromethyl)phenyl]-N′-[(8a,9S)-6′-methoxy-9-cinchonanyl]thiourea

90%

Synonym(s):

1-[3,5-Bis(trifluoromethyl)phenyl)-3-{(S)(6-methoxy-4-quinolinyl)-[(2S,4S,5R)-5-vinyl-1-aza-bicyclo[2.2.2]oct-2-yl]methyl}thiourea, epi-N-Quinyl-N′-bis(3,5-trifluoromethyl)phenylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H28F6N4OS
CAS Number:
Molecular Weight:
594.61
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥89.0%
90%

form

lumps

SMILES string

COc1ccc2nccc([C@H](NC(=S)Nc3cc(cc(c3)C(F)(F)F)C(F)(F)F)C4CC5CCN4C[C@@H]5C=C)c2c1

InChI

1S/C29H28F6N4OS/c1-3-16-15-39-9-7-17(16)10-25(39)26(22-6-8-36-24-5-4-21(40-2)14-23(22)24)38-27(41)37-20-12-18(28(30,31)32)11-19(13-20)29(33,34)35/h3-6,8,11-14,16-17,25-26H,1,7,9-10,15H2,2H3,(H2,37,38,41)/t16-,17-,25-,26-/m0/s1

InChI key

IQMKPBFOEWWDIQ-FRSFCCSCSA-N

Application

N-[3,5-Bis(trifluoromethyl)phenyl]-N′-[(8a,9S)-6′-methoxy-9-cinchonanyl]thiourea is a bifunctional cinchona organocatalyst, which can be used to synthesize:
  • Stereoselective diaryl(nitro)butanone via enantioselective Michael addition of nitromethane to chalcones.
  • Enantioselective β-amino acids via asymmetric Mannich reaction of malonates with aryl and alkyl imines.
  • The synthesis of 3-indolylmethanamines by the reaction of indoles with imines via asymmetric Friedel-Crafts reaction.
  • The enantioselective conjugate addition of active methylene compounds to enones to obtain the corresponding addition products.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Mannich reaction of malonates with simple imines catalyzed by bifunctional cinchona alkaloids: enantioselective synthesis of ?-amino acids
Song J, et al.
Journal of the American Chemical Society, 128(18), 6048-6049 (2006)
Asymmetric Friedel- Crafts reaction of indoles with imines by an organic catalyst
Wang Y-Q, et al.
Journal of the American Chemical Society, 128(25), 8156-8157 (2006)
Organocatalytic enantioselective conjugate additions to enones
Wang J, et al.
Journal of the American Chemical Society, 128(39), 12652-12653 (2006)
Highly enantioselective conjugate addition of nitromethane to chalcones using bifunctional cinchona organocatalysts
Vakulya B, et al.
Organic Letters, 7(10), 1967-1969 (2005)
Urea- and thiourea-substituted cinchona alkaloid derivatives as highly efficient bifunctional organocatalysts for the asymmetric addition of malonate to nitroalkenes: inversion of configuration at C9 dramatically improves catalyst performance.
Séamus H McCooey et al.
Angewandte Chemie (International ed. in English), 44(39), 6367-6370 (2005-09-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service