Skip to Content
Merck
All Photos(1)

Key Documents

659630

Sigma-Aldrich

Polylactide-block-poly(ethylene glycol)-block-polylactide

PLA average Mn 1,500, PEG average Mn 900

Synonym(s):

PLA-PEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CHCH3COO)x(CH2CH2O)y(COCHCH3O)zH
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 900
PLA average Mn 1,500
average Mn (1,500-900-1,500)

degradation timeframe

<12 months

transition temp

Tm 278.5 °C
Tg 7-9 °C

PDI

<1.2

Looking for similar products? Visit Product Comparison Guide

Application

The PLA average Mn value of 3,000 is a combined total between the two PLA blocks.

Features and Benefits

Biodegradable polymer. Applications include drug encapsulation and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service