Skip to Content
Merck
All Photos(1)

Key Documents

616559

Sigma-Aldrich

Fluoxetine-d5 hydrochloride

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D5H13F3NO·HCl
CAS Number:
Molecular Weight:
350.82
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mol wt

350.72 by atom % calculation

mass shift

M+5

SMILES string

Cl.[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(CCNC)Oc2ccc(cc2)C(F)(F)F

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H/i2D,3D,4D,5D,6D;

InChI key

GIYXAJPCNFJEHY-CERKJNTMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenhua Yan et al.
Aquatic toxicology (Amsterdam, Netherlands), 226, 105581-105581 (2020-07-28)
Fluoxetine is a widely prescribed antidepressant that has been frequently detected in aquatic environments and is associated with a series of neurological, behavioural and neuroendocrine disruptions in nontarget organisms. However, studies on its effects in fish under realistic environmental conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service