Skip to Content
Merck
All Photos(1)

Documents

569550

Sigma-Aldrich

Acetonitrile-d3

≥99.8 atom % D, anhydrous

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein:
1740230
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

isotopic purity

≥99.8 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

3.0-16 % (lit.)

technique(s)

NMR: suitable

impurities

>10 ppm water

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

mp

-46 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetonitrile-d3 may be used as:
  • IR and NMR solvent.
  • Probe for zeolite acidity.
  • Cartridge eluent in HPLC– SPE (solid-phase extraction)–NMR technique.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Acetonitrile-d3 as a probe of Lewis and Broensted acidity of zeolites.
Pelmenschikov AG, et al.
The Journal of Physical Chemistry, 97(42), 11071-11074 (1993)
Proton polarizability caused by collective proton motion in intramolecular chains formed by two and three hydrogen bonds. Implications for charge conduction in bacteriorhodopsin.
Brzezinski B, et al.
The Journal of Physical Chemistry, 91(11), 3077-3080 (1987)
Maja Lambert et al.
Magnetic resonance in chemistry : MRC, 43(9), 771-775 (2005-07-29)
The HPLC-SPE-NMR technique was used for the analysis of a root-bark extract of Croton membranaceus. The components of the extract were separated on an analytical-size reversed-phase HPLC column, the chromatographic peaks were trapped on SPE (solid-phase extraction) cartridges after post-column
Dynamics of a wetting liquid in nanopores: An optical Kerr effect study of the dynamics of acetonitrile confined in sol-gel glasses.
Loughnane BJ, et al.
J. Chem. Phys. , 111(11), 5116-5123 (1999)
Organic deuterium compounds: XVI. Synthesis of α-deuterated alkyl nitriles.
Leitch LC.
Canadian Journal of Chemistry, 35(4), 345-347 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service