Skip to Content
Merck
All Photos(1)

Key Documents

535583

Sigma-Aldrich

p-Divinylbenzene

85%

Synonym(s):

1,4-Diethenylbenzene, 1,4-Divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

85%

contains

3000 ppm hydroquinone as stabilizer

composition

meta isomer, ~10 wt. %

refractive index

n20/D 1.5470 (lit.)

density

0.914 g/mL at 25 °C (lit.)

shipped in

dry ice

storage temp.

−70°C

SMILES string

C=Cc1ccc(C=C)cc1

InChI

1S/C10H10/c1-3-9-5-7-10(4-2)8-6-9/h3-8H,1-2H2

InChI key

WEERVPDNCOGWJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Divinylbenzene(DVB) is a cross-linking agent, which is used in a variety of polymeric processes such as copolymerization of styrene to improve the thermo-mechanical properties of the composite.

Application

DVB can be cross-linked with styrene/vinylbenzene chloride and incorporated with multi-walled carbon nanotubes (MWCNTs) to form new microporous nanocomposites for water remediation. It can also be used in combination with polyethylene glycol diacrylate to form linkages with polyethylene oxide (PEO), which can be used as an electrolytic material for lithium batteries.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Linhart et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(6), 645-653 (1989-06-01)
1. Biotransformation of 1,4-diethenylbenzene (1) in rat was studied. Six urinary metabolites, namely, N-acetyl-S-[2-(4-ethenylphenyl)-2-hydroxyethyl]-L-cysteine (3), N-acetyl-S-[1-(4-ethenylphenyl)-2-hydroxyethyl]-L-cysteine (4), N-acetyl-S-[1-(4-formylphenyl)-2-hydroxyethyl]-L-cysteine (5), 1-(4-ethenylphenyl)ethane-1,2-diol (6), 4-ethenylbenzoic acid (9) and 4-ethenylbenzoyl-glycine (12) were isolated and identified by n.m.r. and mass spectrometry. 2. G.l.c.-mass spectral analysis
Novel CeO2 promoted TiO2-ZrO2 nano-oxide catalysts for oxidative dehydrogenation of p-diethylbenzene utilizing CO2 as soft oxidant.
Rao KN, et al.
Applied Catalysis. B, Environmental, 100(3-4), 472-480 (2010)
Monika Wawrzkiewicz et al.
Molecules (Basel, Switzerland), 25(22) (2020-11-15)
Intensive development of many industries, including textile, paper, plastic or food, generate huge amounts of wastewaters containing not only toxic dyes but also harmful auxiliaries such as salts, acid, bases, surfactants, oxidants, heavy metal ions. The search for effective pollutant
Cross-linked solid polymer electrolyte for all-solid-state rechargeable lithium batteries.
Garcia-Calvo O, et al.
Electrochimica Acta, 220(12), 587-594 (2016)
Jakub Hrib et al.
Polymers, 11(7) (2019-06-30)
Self-inflating soft tissue expanders represent a valuable modality in reconstructive surgery. For this purpose, particularly synthetic hydrogels that increase their volume by swelling in aqueous environment are used. The current challenge in the field is to deliver a material with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service