Recommended Products
Assay
98%
mp
81-84 °C (lit.)
SMILES string
COc1ccc2cc(C=O)ccc2c1
InChI
1S/C12H10O2/c1-14-12-5-4-10-6-9(8-13)2-3-11(10)7-12/h2-8H,1H3
InChI key
VZBLASFLFFMMCM-UHFFFAOYSA-N
General description
6-Methoxy-2-naphthaldehyde can be prepared by reacting 2-bromo 6-methoxy naphthalene with triethylorthoformate via Grignard reaction. Its crystals belong to the orthorhombic space group, P212121 and shows excellent NLO (non-linear optical) property.
Application
6-Methoxy-2-naphthaldehyde may be used in the preparation of the following 4-(6-methoxy-2-naphthyl)butan-2-one related compounds:
It may also be used in the synthesis of the following chalcone derivatives:
- 4-(6-methoxy-2-naphthyl)but-3-en-2-one
- 4-(6-methoxy-2-naphthyl)-3-methylbutan-2-one
- 5-(6-methoxy-2-naphthyl)pentan-3-one
It may also be used in the synthesis of the following chalcone derivatives:
- (2E)-1-(2-hydroxyphenyl)-3-(6-methoxy-2-naphthyl)prop-2-en-1-one
- (2E)-1-(2-chloropyridin-4-yl)-3-(6-methoxy-2-naphthyl)prop-2-en-1-one
- (2E)-3-(6-methoxy-2-naphthyl)-1-pyridin-4-ylprop-2-en-1-one
- (2E)-1-(2,4-dichlorophenyl)-3-(6-methoxy-2-naphthyl)prop-2-en-1-one
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cell biochemistry and biophysics, 78(1), 31-42 (2019-11-17)
Human salivary aldehyde dehydrogenase (hsALDH) protects us from the toxic effect of aldehydes. It has both diagnostic and therapeutic importance. Citral possesses many biological and pharmacological properties. The aim of this work was to investigate the inhibitory effect and the
Acta poloniae pharmaceutica, 53(3), 203-208 (1996-05-01)
Continuous fluorimetric assay for aldehyde dehydrogenase activity in rat tissue, based on oxidation of the fluorogenic 6-methoxy-2-naphthaldehyde, is described. The new assay is ca. 2 orders of magnitude more sensitive than the standard procedures and it is not subjected to
Growth characterization and non linear optical property of 6-methoxy-2-naphthaldehyde.
Proceedings of SPIE, 5935 (2005)
4-(6-Methoxy-2-naphthyl) butan-2-one and related analogs, a novel structural class of antiinflammatory compounds.
Journal of Medicinal Chemistry, 21(12), 1260-1264 (1978)
Chemosphere, 243, 125358-125358 (2019-11-24)
Human salivary aldehyde dehydrogenase (hsALDH) is an important detoxifying enzyme and maintains oral health. Subjects with low hsALDH activity are at a risk of developing oral cancers. Arsenic (As) toxicity causes many health problems in humans. The objective of this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service