Skip to Content
Merck
All Photos(1)

Key Documents

492655

Sigma-Aldrich

Formamide-1-d

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCONH2
CAS Number:
Molecular Weight:
46.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.159 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]C(N)=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)/i1D

InChI key

ZHNUHDYFZUAESO-MICDWDOJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

347.0 °F

Flash Point(C)

175 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service