Skip to Content
Merck
All Photos(1)

Documents

464481

Sigma-Aldrich

2-Octyl-1-dodecanol

97%

Synonym(s):

Octyldodecanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH[(CH2)7CH3]CH2OH
CAS Number:
Molecular Weight:
298.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.453 (lit.)

bp

234-238 °C/33 mmHg (lit.)

mp

−1-1 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCC(CO)CCCCCCCC

InChI

1S/C20H42O/c1-3-5-7-9-11-12-14-16-18-20(19-21)17-15-13-10-8-6-4-2/h20-21H,3-19H2,1-2H3

InChI key

LEACJMVNYZDSKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Octyl-1-dodecanol is a branched alcohol.

Application

2-Octyl-1-dodecanol may be used to investigate its interaction with the hexameric capsules of resorcin[4]arene. It has been used as diluent in a extractant screening study for the recovery of putrescine (butylene-1,4-diamine, BDA) and cadaverine (pentylene-1,5-diamine, PDA) from aqueous solutions (fermentation broths) by liquid-liquid extraction.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

370.4 °F - open cup

Flash Point(C)

188 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy Sharma et al.
Artificial cells, nanomedicine, and biotechnology, 45(3), 409-413 (2016-03-26)
The objective of present work was to enquire the potential use of embelin-loaded nanolipid carriers for brain targeting. The average particle size and polydispersity index (PDI) of optimized formulation (F19) were found to be 152 ± 19.7 nm and 0.143 ± 0.023, respectively. Nanolipid carrier
Extractive recovery of aqueous diamines for bio-based plastics production.
Krzyzaniak A, et al.
Journal of Chemical Technology and Biotechnology, 88(10), 1937-1945 (2013)
Contact sensitivity to octyldodecanol and trometamol in an anti-itch cream.
M Singh et al.
Contact dermatitis, 56(5), 289-290 (2007-04-20)
R S Lanigan
International journal of toxicology, 20 Suppl 3, 51-59 (2002-01-05)
Octyldodecyl Stearoyl Stearate functions as an occlusive skin-conditioning agent and as a nonaqueous viscosity-increasing agent in many cosmetic formulations. Current concentrations of use are between 0.7% and 23%, although historically higher concentrations were used. The chemical is formed by a
Strontium removal by new alkyl phenylphosphonic acids in supported liquid membranes with strip dispersion.
Ho WSW and Wang W.
Industrial & Engineering Chemistry Research, 41(3), 381-388 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service