Skip to Content
Merck
All Photos(2)

Key Documents

459003

Sigma-Aldrich

Polyethylene-block-poly(ethylene glycol)

average Mn ~575

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2(CH2CH2)X(OCH2CH2)YOH
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mol wt

average Mn ~575

composition

ethylene oxide, 20 wt. %

hydroxyl value

85 mg KOH/g

transition temp

Tm 101 °C (DSC)

solubility

mineral oil, cyclohexane, toluene and MEK: soluble

HLB

4

SMILES string

C=C.OCCO

Looking for similar products? Visit Product Comparison Guide

Application

Nonionic surfactant.
Emulsifier for coatings, ceramics, metalworking fluids and inks. Lubricant. Mold release agent. Thickening agent.
Nonionic surfactant.

Features and Benefits

Polyether segment is susceptible to oxidative degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service