Skip to Content
Merck
All Photos(3)

Documents

449458

Sigma-Aldrich

(tert-Butyldimethylsilyloxy)acetaldehyde

90%

Synonym(s):

(tert-Butyldimethylsiloxy)acetaldehyde, 2-(tert-Butyldimethylsilyloxy)acetaldehyde, 2-[(tert-Butyl)dimethylsiloxy]acetaldehyde, 2-[(tert-Butyldimethylsilanyl)oxy]acetaldehyde, 2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]acetaldehyde, Dimethyl-tert-butylsilyloxyacetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2OCH2CHO
CAS Number:
Molecular Weight:
174.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.432 (lit.)

bp

165-167 °C (lit.)

density

0.915 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OCC=O

InChI

1S/C8H18O2Si/c1-8(2,3)11(4,5)10-7-6-9/h6H,7H2,1-5H3

InChI key

MEBFFOKESLAUSJ-UHFFFAOYSA-N

Application

(tert-Butyldimethylsilyloxy)acetaldehyde is a versatile reagent commonly used in synthetic glycobiology. It can act both as an aldol donor and an aldol acceptor in the stereocontrolled production of erythrose. It is used as an important reagent in the total synthesis of (+)-ambruticin, (−)-laulimalide, (−)-salinosporamide A, and (+)-leucascandrolide A.
Employed in the construction of the key tetrahydropyran subunit in a recent synthesis of the marine natural product (–)-dactylodide.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Biological Evaluation of the Trifluoromethyl Analog of (4S)-4, 5-Dihydroxy-2, 3-pentanedione (DPD).
Frezza M, et al.
European Journal of Organic Chemistry, 20, 4731-4736 (2006)
Total synthesis of (+)-ambruticin.
Liu P and Jacobsen EN
Journal of the American Chemical Society, 123(43), 10772-10773 (2001)
Colobert, Francoise; et al.
European Journal of Organic Chemistry, 6, 1455-1467 (2006)
Total synthesis of the microtubule-stabilizing agent (−)-Laulimalide
Paterson I, et al.
Organic Letters, 3(20), 3149-3152 (2001)
Synthesis of (?)-thioascorbic acid.
Stachel H-D, et al.
Tetrahedron, 49(22), 4871-4880 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service