Skip to Content
Merck
All Photos(1)

Documents

425508

Sigma-Aldrich

2,6-Dimethylphenylmagnesium bromide solution

1.0 M in THF

Synonym(s):

2,6-Xylylmagnesium bromide, Bromo(2,6-dimethylphenyl)magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3MgBr
CAS Number:
Molecular Weight:
209.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

bp

65-67 °C

density

0.998 g/mL at 25 °C

SMILES string

Cc1cccc(C)c1[Mg]Br

InChI

1S/C8H9.BrH.Mg/c1-7-4-3-5-8(2)6-7;;/h3-5H,1-2H3;1H;/q;;+1/p-1

InChI key

BIGUVDUZGAJWQK-UHFFFAOYSA-M

Application

2,6-Dimethylphenylmagnesium bromide can be used:
  • As an intermediate in the synthesis of dibutyl(5,10-dihydro-1,9-di-p-toluoyl-5-p-tolyldipyrrinato)tin(IV), which is further used in the preparation of zinc-based porphyrin.
  • As a substrate in the preparation of substituted fluorenes by reacting with 2-substituted arenesulfonates and dihaloarenes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd (OAc) 2-catalyzed domino reactions of 1, 2-dihaloarenes and 2-haloaryl arenesulfonates with Grignard reagents: efficient synthesis of substituted fluorenes
Dong C and Hu Q
Tetrahedron, 64(11), 2537-2552 (2008)
Nearly Chromatography-Free Synthesis of the A3B-Porphyrin 5-(4-Hydroxymethylphenyl)-10, 15, 20-tri-p-tolylporphinatozinc (II).
Zaidi SHH, et al.
Organic Process Research & Development, 10(2), 304-314 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service