Skip to Content
Merck
All Photos(1)

Key Documents

362530

Sigma-Aldrich

Aluminum phthalocyanine chloride

Dye content ~85 %

Synonym(s):

Chloro(29H,31H-phthalocyaninato)aluminum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16AlClN8
CAS Number:
Molecular Weight:
574.96
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, ~85%

mp

>300 °C (lit.)

λmax

680 nm

SMILES string

Cl[Al]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Al.ClH/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;;1H/q-2;+3;/p-1

InChI key

LBGCRGLFTKVXDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela B Rodrigues et al.
Photochemistry and photobiology, 88(2), 440-447 (2011-12-08)
The selection of fungi resistant to currently used fungicides and the emergence of new pathogenic species make the development of alternative fungus-control techniques highly desirable. Photodynamic antimicrobial chemotherapy (PACT) is a promising method which combines a nontoxic photosensitizer (PS) with
E C C Tapajós et al.
Oral oncology, 44(11), 1073-1079 (2008-07-16)
In this study, oral carcinoma cells were used to evaluate chloroaluminum-phthalocyanine encapsulated in liposomes as the photosensitizer agent in support of photodynamic therapy (PDT). The genotoxicity and cytotoxicity behavior of the encapsulated photosensitizer in both dark and under irradiation using
Daniela Cervelle Zancanela et al.
Photomedicine and laser surgery, 29(10), 699-705 (2011-06-15)
A promising new treatment in dentistry involves the photodynamic process, which utilizes a combination of two therapeutic agents, namely a photosensitizer drug and a low dose of visible light. We investigated the in vitro effect of low intensity laser irradiation
Leandro Santos Bicalho et al.
Journal of biomedical nanotechnology, 9(5), 811-818 (2013-06-28)
In patients diagnosed with oral cancer, the most important prognostic indicator for patient survival after primary treatment is metastasis to the cervical lymph nodes or distal sites. Therefore, we evaluated the utility of photodynamic therapy (PDT) mediated by aluminum-chloride-phthalocyanine entrapped
Shuai Zhang et al.
Molecules (Basel, Switzerland), 17(9), 10801-10815 (2012-09-12)
Photoelectrochemical measurements were used to study the photoelectrode characteristics of composite nanoparticles composed of fullerene C₆₀ and partially hydrolyzed aluminum phthalocyanine chloride (AlPc). In cyclic voltammetry measurements, the electrodes coated with the composite nanoparticles were found to have photoanodic [electron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service