Skip to Content
Merck
All Photos(1)

Documents

331597

Sigma-Aldrich

2-Methoxythiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

151-152 °C/762 mmHg (lit.)

density

1.133 g/mL at 25 °C (lit.)

SMILES string

COc1cccs1

InChI

1S/C5H6OS/c1-6-5-3-2-4-7-5/h2-4H,1H3

InChI key

OKEHURCMYKPVFW-UHFFFAOYSA-N

General description

2-Methoxythiophene is a heterocyclic methyl enol ether and its reaction with o-quinone monoimide was studied. The intramolecular and intermolecular geometries of crystals of 2-methoxythiophene were investigated. Kinetics of the hydronium-ion catalysed hydrolysis of 2-methoxythiophene was reported.

Application

2-Methoxythiophene was used in thermal reaction (60°C) of (C5Me5)Rh(PMe3)(Ph)H.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of an o-quinone monoimide with 1, 3, 5-trimethoxybenzene, 2-methoxythiophene, 2-methoxyfuran, and 1-methyl-, 2-methyl-, and 1, 2-dimethylindoles.
Heine HW, et al.
The Journal of Organic Chemistry, 52(1), 97-101 (1987)
Reversible carbon protonation in the hydrolysis of heterocyclic enol methyl ethers.
Capon B and Kwok F-C.
Tetrahedron, 43(1), 69-76 (1987)
Bond cleavage reactions in substituted thiophenes by a rhodium complex.
Myers AW, et al.
Inorgorganica Chimica Acta, 361(11), 3263-3270 (2008)
Blake et al.
Acta crystallographica. Section B, Structural science, 55(Pt 6), 963-974 (2000-08-06)
The intramolecular and intermolecular geometries of six thiophenes carrying oxygen-containing substituents have been determined. Crystals of 2-methoxythiophene and 3-methoxythiophene were grown in situ on a diffractometer from liquid samples. The 2-methoxy group introduces significant distortions to the thiophene nucleus and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service